Learn More
Cytiva ExoProStar DNA and Sequence Reaction Clean-up
Optimized to purify DNA and sequencing set up reactions quickly, efficiently and reliably. illustra™ ExoProStar DNA and Sequence Reaction Clean-up combines enzymatic PCR clean up technology with illustra Exonuclease I and Alkaline Phosphatase to remove unincorporated primers and dNTPs.
Brand:  Cytiva US78210
67.97 GBP valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Code : GH
Additional Details : Weight : 0.08000kg
 View more versions of this product
Product Code. 11663044
  • £73.88 / Each
Estimated Shipment: 30-05-2024
to see stock.
Description
Description
Enzymes optimized to work together for high efficiency removal of unincorporated primers and dNTPs. Enzymes provided in two separate tubes, just two simple pipetting steps are needed to prepare the reaction.
  • Fast 30-minute protocol
  • Scalable for different reaction sizes
  • No loss of PCR product
  • Easy to automate
  • Complete heat inactivation of the enzymes within 15 minutes
Specifications
Specifications
Enzymatic PCR clean up technology with illustra Exonuclease I and Alkaline Phosphatase to remove unincorporated primers and dNTPs. For maximum flexibility comes in two separate tubes.
One tube of Alkaline Phosphatase and Exonuclease I
illustra ExoProStar
Optimized to purify DNA and sequencing set up reactions quickly, efficiently and reliably
100