missing translation for 'onlineSavingsMsg'
Learn More

ILKAP/PP2C delta Antibody, DyLight 594, Novus Biologicals™

Product Code. 18396863 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18396863 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18396863 Supplier Novus Biologicals Supplier No. NBP300036DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ILKAP/PP2C delta Polyclonal specifically detects ILKAP/PP2C delta in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ILKAP/PP2C delta
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias DKFZp434J2031, EC 3.1.3.16, FLJ10181, integrin-linked kinase-associated serine/threonine phosphatase, integrin-linked kinase-associated serine/threonine phosphatase 2C, MGC4846, PP2C-DELTA, protein phosphatase 2c, delta isozyme
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant ILKAP/PP2C delta (Q9H0C8; Met1-His392).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 80895
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.