missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ILK Recombinant Rabbit Monoclonal Antibody (6A5)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA549818
This item is not returnable.
View return policy
Description
ILK Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IP, ELISA
Transduction of extracellular matrix signals through integrins influences intracellular and extracellular functions, and appears to require interaction of integrin cytoplasmic domains with cellular proteins. Integrin-linked kinase (ILK), interacts with the cytoplasmic domain of beta-1 integrin. This gene encodes a serine/threonine protein kinase with 4 ankyrin-like repeats, which associates with the cytoplasmic domain of beta integrins and acts as a proximal receptor kinase regulating integrin-mediated signal transduction. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
| ILK | |
| Recombinant Monoclonal | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O55222, Q13418 | |
| ILK | |
| A synthesized peptide derived from human ILK. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 6A5 | |
| Unconjugated | |
| ILK | |
| 59 kDa serine/threonine-protein kinase; AA511515; epididymis secretory protein Li 28; ESTM24; HEL-S-28; ILK; ILK1; ILK-1; ILK2; ILK-2; integrin binding protein kinase; Integrin linked ILK; integrin linked kinase; integrin-linked kinase; integrin-linked kinase-2; integrin-linked protein kinase; loc; lost-contact; main squeeze; msq; P59; p59ILK; zgc:65842; zilk | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 16202, 3611 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction