missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ILK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119671
This item is not returnable.
View return policy
Description
Positive Control: Mouse skeletal muscle tissue, rat lung tissue, A431, MG-63, PANC-1, rat skeletal muscle tissue, human kidney tissue, human pancreas tissue, mouse heart tissue. Subcellular Location: Cell junction, Cell membrane, Cytoplasm.
Transduction of extracellular matrix signals through integrins influences intracellular and extracellular functions, and appears to require interaction of integrin cytoplasmic domains with cellular proteins. Integrin-linked kinase (ILK), interacts with the cytoplasmic domain of beta-1 integrin. This gene encodes a serine/threonine protein kinase with 4 ankyrin-like repeats, which associates with the cytoplasmic domain of beta integrins and acts as a proximal receptor kinase regulating integrin-mediated signal transduction. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
| ILK | |
| Polyclonal | |
| Unconjugated | |
| ILK | |
| 59 kDa serine/threonine-protein kinase; AA511515; epididymis secretory protein Li 28; ESTM24; HEL-S-28; ILK; ILK1; ILK-1; ILK2; ILK-2; integrin binding protein kinase; Integrin linked ILK; integrin linked kinase; integrin-linked kinase; integrin-linked kinase-2; integrin-linked protein kinase; loc; lost-contact; main squeeze; msq; P59; p59ILK; zgc:65842; zilk | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16202, 170922, 3611 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| O55222, Q13418, Q99J82 | |
| ILK | |
| Recombinant protein within Human Integrin linked ILK aa 1-290. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu