missing translation for 'onlineSavingsMsg'
Learn More

ILK-IN-2, MedChemExpress

Product Code. 16410058
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
Unit Size:
10mg
2mg
5mg
This item is not returnable. View return policy

Product Code. 16410058

Brand: Medchem Express HY18676B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ILK-IN-2 (OSU-T315 analog) is a ILK inhibitor.

ILK-IN-2 (OSU-T315 analog) is a ILK inhibitor.

Specifications

Chemical Name or Material ILK-IN-2
Physical Form Solid
CAS 1333146-24-9
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C30H30F3N5O
For Use With (Application) Cancer-programmed cell death
Synonym OSU-T315 analog
Solubility Information DMSO : 50 mg/mL (93.70 mM; Need ultrasonic)
SMILES O=C(NC)CCC1=NN(C2=CC=C(N3CCNCC3)C=C2)C(C4=CC=C(C5=CC=C(C(F)(F)F)C=C5)C=C4)=C1
Molecular Weight (g/mol) 533.59
Formula Weight 533.59
Percent Purity 98.14%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.