missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILK Antibody (OTI2H3), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71037APC
This item is not returnable.
View return policy
Description
ILK Monoclonal antibody specifically detects ILK in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| ILK | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| OTI2H3 | |
| Western Blot | |
| 59 kDa serine/threonine-protein kinase, DKFZp686F1765, EC 2.7.11.1, ILK1, ILK-1, ILK2, ILK-2, integrin-linked kinase, integrin-linked kinase-2, integrin-linked protein kinase, P59, p59ILK | |
| Human recombinant protein fragment corresponding to amino acids 1-244 of human ILK (NP_004508) produced in E.coli. | |
| 0.1 mL | |
| Phospho Specific, Protein Kinase, Signal Transduction | |
| 3611 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction