missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILK Antibody (6A5), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | ILK |
|---|---|
| Clone | 6A5 |
| Dilution | Western Blot 1:500-1:5000, ELISA, Immunoprecipitation 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
| Applications | Western Blot, ELISA, Immunoprecipitation, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30039977
|
Novus Biologicals
NBP3-26198-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30027965
|
Novus Biologicals
NBP3-26198-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ILK Monoclonal antibody specifically detects ILK in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunoprecipitation, ImmunocytochemistrySpecifications
| ILK | |
| Western Blot 1:500-1:5000, ELISA, Immunoprecipitation 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| 59 kDa serine/threonine-protein kinase, DKFZp686F1765, EC 2.7.11.1, ILK1, ILK-1, ILK2, ILK-2, integrin-linked kinase, integrin-linked kinase-2, integrin-linked protein kinase, P59, p59ILK | |
| A synthesized peptide derived from Human ILK [UniProt Q13418] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 6A5 | |
| Western Blot, ELISA, Immunoprecipitation, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Phospho Specific, Protein Kinase, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 3611 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title