missing translation for 'onlineSavingsMsg'
Learn More

Ilginatinib, MedChemExpress

Product Code. 16493777
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493777 50 mg 50mg
16457828 10 mg 10mg
16437656 100 mg 100mg
16400298 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16493777 Supplier Medchem Express Supplier No. HY19631A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ilginatinib (NS-018) is a highly active and orally bioavailable JAK2 inhibitor, with an IC50 of 0.72 nM, 46-, 54-, and 31-fold selectivity for JAK2 over JAK1 (IC50, 33 nM), JAK3 (IC50, 39 nM), and Tyk2 (IC50, 22 nM).

Ilginatinib (NS-018) is a highly active and orally bioavailable JAK2 inhibitor, with an IC50 of 0.72 nM, 46-, 54-, and 31-fold selectivity for JAK2 over JAK1 (IC50, 33 nM), JAK3 (IC50, 39 nM), and Tyk2 (IC50, 22 nM).

Specifications

Chemical Name or Material Ilginatinib
CAS 1239358-86-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H20FN7
Quantity 50 mg
Synonym NS-018
Solubility Information DMSO : 100 mg/mL (256.79 mM; Need ultrasonic)
SMILES CN1N=CC(C2=CC(N[C@H](C3=CC=C(F)C=C3)C)=NC(NC4=NC=CN=C4)=C2)=C1
Molecular Weight (g/mol) 389.43
Formula Weight 389.43
Percent Purity 95.07%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.