missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ilginatinib maleate, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
Ilginatinib maleate (NS-018 maleate) is a highly active and orally bioavailable JAK2 inhibitor, with an IC50 of 0.72 nM, 46-, 54-, and 31-fold selectivity for JAK2 over JAK1 (IC50, 33 nM), JAK3 (IC50, 39 nM), and Tyk2 (IC50, 22 nM).
Specifications
Specifications
| Chemical Name or Material | Ilginatinib maleate |
| CAS | 1354799-87-3 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C25H24FN7O4 |
| Quantity | 5 mg |
| Synonym | NS-018 maleate |
| Solubility Information | DMSO : ≥ 30 mg/mL (59.35 mM) ∣H2O : 1 mg/mL (1.98 mM; Need ultrasonic) |
| SMILES | CN1N=CC(C2=CC(N[C@H](C3=CC=C(F)C=C3)C)=NC(NC4=NC=CN=C4)=C2)=C1.O=C(O)/C=C\C(O)=O |
| Molecular Weight (g/mol) | 505.5 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction