missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILF3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19887-1-AP-20UL
This item is not returnable.
View return policy
Description
UBA6 (UBE1L2, MOP-4)is a novel E1 enzyme specific for ubiquitin and belongs to the ubiquitin-activating E1 family. It is a part of the cascade to attach ubiquitin (Ub) covalently to substrate proteins.UBA6 activates ubiquitin by first adenylating its carboxy-terminal glycine residue with ATP and then linking this residue to the side chain of a cysteine residue in E1, yielding an ubiquitin-E1 thioester and free AMP. Its isoforms influence spermatogenesis and male fertility.UBA6 is highly expressed in human monocytes.Specifications
| ILF3 | |
| Polyclonal | |
| Unconjugated | |
| ILF3 | |
| CBTF, DRBF, DRBP76, ILF3, M phase phosphoprotein 4, MMP4, MPHOSPH4, MPP4, NF AT 90, NF110, NF90, NFAR, NFAR 1, NFAR2, TCP110, TCP80 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16201, 3609, 84472 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q12906, Q9JIL3, Q9Z1X4 | |
| ILF3 | |
| ILF3 Fusion Protein Ag13707 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction