missing translation for 'onlineSavingsMsg'
Learn More

ILF3 Rabbit anti-Human, Mouse, Rat, Clone: 0S2N7, Novus Biologicals™

Product Code. 18344344 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18344344 100 μg 100µL
18314945 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18344344 Supplier Novus Biologicals Supplier No. NBP316664100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ILF3 Monoclonal antibody specifically detects ILF3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ILF3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 0S2N7
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Double-stranded RNA-binding protein 76, double-stranded RNA-binding protein, 76 kD, DRBF, DRBP76M-phase phosphoprotein 4, dsRNA binding protein NFAR-2/MPP4, interleukin enhancer binding factor 3, 90kDa, interleukin enhancer-binding factor 3, MPHOSPH4interleukin enhancer binding factor 3, 90kD, MPP4MMP4, NF90CBTF, NFAR-1, NFAR2, NFARNF110, NF-AT-90, Nuclear factor associated with dsRNA, Nuclear factor of activated T-cells 90 kDa, nuclear factor of activated T-cells, 90 kD, TCP110, TCP80, Translational control protein 80
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ILF3 (Q12906).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 3609
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.