missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILF3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-58225
This item is not returnable.
View return policy
Description
ILF3 Polyclonal specifically detects ILF3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ILF3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Double-stranded RNA-binding protein 76, double-stranded RNA-binding protein, 76 kD, DRBF, DRBP76M-phase phosphoprotein 4, dsRNA binding protein NFAR-2/MPP4, interleukin enhancer binding factor 3, 90kDa, interleukin enhancer-binding factor 3, MPHOSPH4interleukin enhancer binding factor 3, 90kD, MPP4MMP4, NF90CBTF, NFAR-1, NFAR2, NFARNF110, NF-AT-90, Nuclear factor associated with dsRNA, Nuclear factor of activated T-cells 90 kDa, nuclear factor of activated T-cells, 90 kD, TCP110, TCP80, Translational control protein 80 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Guinea pig: 100%; Human: 100%; Equine: 92%; Bovine: 85%; Canine: 85%; Rabbit: 84%. | |
| Human, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 4-8 ug/ml | |
| Q12906-2 | |
| ILF3 | |
| Synthetic peptides corresponding to ILF3 (interleukin enhancer binding factor 3, 90kDa) The peptide sequence was selected from the N terminal of ILF3 (NP_004507). Peptide sequence PTQEELEAVQNMVSHTERALKAVSDWIDEQEKGSSEQAESDNMDVPPEDD. | |
| 100 μL | |
| Cell Cycle and Replication | |
| 3609 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction