missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILF3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-37999-100ul
This item is not returnable.
View return policy
Description
ILF3 Polyclonal antibody specifically detects ILF3 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| ILF3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Double-stranded RNA-binding protein 76, double-stranded RNA-binding protein, 76 kD, DRBF, DRBP76M-phase phosphoprotein 4, dsRNA binding protein NFAR-2/MPP4, interleukin enhancer binding factor 3, 90kDa, interleukin enhancer-binding factor 3, MPHOSPH4interleukin enhancer binding factor 3, 90kD, MPP4MMP4, NF90CBTF, NFAR-1, NFAR2, NFARNF110, NF-AT-90, Nuclear factor associated with dsRNA, Nuclear factor of activated T-cells 90 kDa, nuclear factor of activated T-cells, 90 kD, TCP110, TCP80, Translational control protein 80 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human ILF3 (NP_036350.2).,, Sequence:, MRPMRIFVNDDRHVMAKHSSVYPTQEELEAVQNMVSHTERALKAVSDWIDEQEKGSSEQAESDNMDVPPEDDSKEGAGEQKTEHMTRTLRGVMRVGLVAKGLLLKGDLDLELVLLCKEKPTTALLDKVADNLAIQLAAVTEDKYEILQSVDDAAIVIKNTKEPPLSLTIHLTSPVVREEMEKVLAGETLSVNDPPDVLDRQKCLAALASLRHAKWFQARANGLKSCVIVIRVLRDLCTRVPTWGPLRGWPLELLCEKSIGTANRPMGAGE | |
| 100 μL | |
| Cell Cycle and Replication | |
| 3609 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction