missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-88147
This item is not returnable.
View return policy
Description
ILF1 Polyclonal specifically detects ILF1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| ILF1 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| Cellular transcription factor ILF-1, forkhead box K2, forkhead box protein K2, FOXK1, ILF-1, ILF1ILF, interleukin enhancer binding factor 1, Interleukin enhancer-binding factor 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| NP_004505.2 | |
| FOXK2 | |
| The immunogen recognized by this antibody maps to a region between residue 600 and 635 of human forkhead box K2 using the numbering given in entry NP_004505.2 (GeneID 3607). | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 3607 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction