missing translation for 'onlineSavingsMsg'
Learn More

Ilaprazole, TRC

Product Code. 30396334
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396334 2.5 mg 2.5mg
30404974 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30396334 Supplier Toronto Research Chemicals Supplier No. TRCI2664002.5MG
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 172152-36-2
Molecular Formula C19 H18 N4 O2 S
Molecular Weight (g/mol) 366.44
Synonym 2-[[(4-Methoxy-3-methyl-2-pyridinyl)methyl]sulfinyl]-6-(1H-pyrrol-1-yl)-1H-benzimidazole, 1H-Benzimidazole, 2-[[(4-methoxy-3-methyl-2-pyridinyl)methyl]sulfinyl]-5-(1H-pyrrol-1-yl)- (9CI), 2-[[(4-Methoxy-3-methyl-2-pyridinyl)methyl]sulfinyl]-6-(1H-pyrrol-1-yl)-1H-benzimidazole, IY 81149, Ilaprazole, 2-[[(4-Methoxy-3-methyl-2-pyridinyl)methyl]sulfinyl]-6-(1H-pyrrol-1-yl)-1H-benzimidazole, 1H-Benzimidazole, 2-[[(4-methoxy-3-methyl-2-pyridinyl)methyl]sulfinyl]-5-(1H-pyrrol-1-yl)- (9CI), 2-[[(4-Methoxy-3-methyl-2-pyridinyl)methyl]sulfinyl]-6-(1H-pyrrol-1-yl)-1H-benzimidazole, IY 81149
IUPAC Name 2-[(4-methoxy-3-methylpyridin-2-yl)methylsulfinyl]-5-pyrrol-1-yl-1H-benzimidazole
SMILES COc1ccnc(CS(=O)c2nc3cc(ccc3[nH]2)n4cccc4)c1C

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C19H18N4O2S/c1-13-17(20-8-7-18(13)25-2)12-26(24)19-21-15-6-5-14(11-16(15)22-19)23-9-3-4-10-23/h3-11H,12H2,1-2H3,(H,21,22)
Formula Weight 366.115
Chemical Name or Material Ilaprazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.