missing translation for 'onlineSavingsMsg'
Learn More

Ilaprazole, MedChemExpress

Product Code. 16494656
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494656 100 mg 100mg
16479956 50 mg 50mg
16470056 2 mg 2mg
16463488 10 mg 10mg
16416217 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16494656 Supplier Medchem Express Supplier No. HY101664
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ilaprazole (IY-81149) is an orally active proton pump inhibitor. Ilaprazole irreversibly inhibits H+/K+-ATPase in a dose-dependent manner with an IC50 of pump inhibitory activity of 6 μM in rabbit parietal cell preparation. Ilaprazole is used for the research of gastric ulcers. Ilaprazole is also a potent TOPK (T-lymphokine-activated killer cell-originated protein kinase) inhibitor.

Ilaprazole (IY-81149) is an orally active proton pump inhibitor. Ilaprazole irreversibly inhibits H+/K+-ATPase in a dose-dependent manner with an IC50 of pump inhibitory activity of 6 μM in rabbit parietal cell preparation. Ilaprazole is used for the research of gastric ulcers. Ilaprazole is also a potent TOPK (T-lymphokine-activated killer cell-originated protein kinase) inhibitor.

Specifications

Chemical Name or Material Ilaprazole
CAS 172152-36-2
Color Light Nude
Purity Grade Notes Research
Molecular Formula C19H18N4O2S
Quantity 100 mg
Synonym IY-81149
Solubility Information DMSO : ≥ 35 mg/mL (95.51 mM) ∣Ethanol : 12.5 mg/mL (34.11 mM; Need ultrasonic)
SMILES O=S(C1=NC2=CC=C(N3C=CC=C3)C=C2N1)CC4=NC=CC(OC)=C4C
Molecular Weight (g/mol) 366.44
Formula Weight 366.44
Percent Purity 99.26%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.