missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL9R Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148927
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Human Interleukin 9 (IL9) was first identified based on its proliferative activity on a human myeloid leukemic cell line. Human IL9 stimulates the proliferation of the human megakaryocytic leukemic cell line M07e. Human IL9 supports erythroid colony formation and synergizes with IL4 in the production of IgE and IgG. The interleukin 9 receptor is a member of the hematopoietin receptor superfamily. The cDNAs encoding mouse and human IL9 receptors have been isolated. The deduced mouse and human transmembrane proteins, sharing 53% amino acid sequence identity, contain 468 and 533 amino acid residues, respectively. A number of isoforms of IL9R, including a putative soluble form, have also been identified. The IL9 receptor is expressed in a variety of hematopoietic cells including T cells, neutrophils, mast cells, and macrophages.
Specifications
| IL9R | |
| Polyclonal | |
| Unconjugated | |
| IL9R | |
| IL9R | |
| Mouse myeloma cell line NS0-derived recombinant rat IL-9 R, Val38-Ala270. | |
| 100 μg | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| CD129; IL-9 receptor; Il9r; IL-9R; interleukin 9 receptor; interleukin-9 receptor | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 24500 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction