missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sino Biological™ IL8 Recombinant Human Protein
Click to view available options
Quantity:
5 x 5 μg
Unit Size:
25µg
Description
Expressed from DNA sequence encoding 77 amino acid endothelial-cell derived form of mature human IL8 (Ala 23-Ser 99) fused to the Fc region of human IgG1
- 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80
Specifications
Specifications
| For Use With (Application) | Assay Standard |
| Gene ID (Entrez) | 3576 |
| Molecular Weight (g/mol) | 35.6kd |
| Quantity | 5 x 5 μg |
| Source | Human |
| Storage Requirements | −20°C |
| Structural Form | Truncated, Recombinant |
| Endotoxin Concentration | <1 EU/μg |
| Common Name | Interleukin 8 |
| Gene Symbol | IL8 |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction