missing translation for 'onlineSavingsMsg'
Learn More

IL6R Mouse anti-Human, Clone: B-F19, Novus Biologicals™

Product Code. 18368166
Change view
Click to view available options
Quantity:
200 μg
500 μg
Unit Size:
200µL
500µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368166 500 μg 500µL
18309151 200 μg 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18368166 Supplier Novus Biologicals Supplier No. NBP318095500UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL6R Monoclonal antibody specifically detects IL6R in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL6R
Applications Flow Cytometry, ELISA, Immunohistochemistry
Classification Monoclonal
Clone B-F19
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry, ELISA, Immunohistochemistry
Formulation PBS. Sterile-filtered through 0.22 um.
Gene Alias CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991
Host Species Mouse
Immunogen gp80 transfected CHO cell line. Hybridoma: Myeloma X63/AG.8653 x Balb/c spleen cells
Purification Method Affinity purified
Quantity 500 μg
Regulatory Status RUO
Research Discipline Cancer, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3570
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.