missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IL6R Monoclonal specifically detects IL6R in Human samples. It is validated for ELISA, Immunofluorescence.
Specifications
Specifications
| Antigen | IL6R |
| Applications | ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | rhPM-1 (Tocilizumab) |
| Conjugate | DyLight 755 |
| Dilution | ELISA, Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991 |
| Host Species | Human |
| Immunogen | This antibody was prepared by CDR grafting of mouse AUK 12-20 antibody onto a human framework regions. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?