missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL6R Antibody (AP-MAB0848) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-06670-0.1mg
This item is not returnable.
View return policy
Description
IL6R Monoclonal specifically detects IL6R in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, CyTOF-ready, Block/Neutralize.
Specifications
| IL6R | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500, Blocking/Neutralizing 50-200 molar excess, CyTOF-ready | |
| P22272 | |
| IL6R | |
| Mouse recombinant IL-6R alpha chain | |
| 0.1 mg | |
| Cancer, Immunology, Innate Immunity | |
| 3570 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles. | |
| IgG2 |
| Flow Cytometry | |
| AP-MAB0848 | |
| Unconjugated | |
| PBS with No Preservative | |
| CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991 | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction