missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL5RA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581422
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Mouse IL5RA (CD125).
Interleukin 5 (IL-5) is a soluble T cell-derived factor, also known as T cell replacing factor (TRF), that induces B cell and eosinophil growth and differentiation. IL-5 exerts its biological activity through the IL-5 receptor (IL-5R), which is composed of two chains: an alpha chain that binds IL-5 with low affinity and a beta chain that does not bind IL-5, but together with the IL-5 alpha chain, constitutes the high affinity IL-5 receptor. The cytoplasmic domain of both the alpha and beta chains is essential for signal transduction. Specifically, the membrane-proximal proline-rich sequence of the cytoplasmic domain of the IL-5R receptor alpha subunit, IL-5Ralpha, is critical for the IL-5 induced proliferative response, expression of nuclear proto-oncogenes and tyrosine phosphorylation of cellular proteins, such as JAK1 and JAK2.
Specifications
| IL5RA | |
| Polyclonal | |
| Unconjugated | |
| IL5RA | |
| CD125; CD125 antigen; CDw125; HSIL5R3; IL-5 receptor alpha chain; IL-5 receptor subunit alpha; Il5r; IL-5R subunit alpha; Il5ra; IL-5RA; IL-5R-alpha; interleukin 5 receptor subunit alpha; interleukin 5 receptor type 3; interleukin 5 receptor, alpha; interleukin-5 receptor alpha chain; interleukin-5 receptor subunit alpha | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 16192 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| P21183 | |
| IL5RA | |
| Recombinant Mouse IL5RA (CD125) Protein (Met1-Val328). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction