missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL4R Recombinant Rabbit Monoclonal Antibody (102), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£124.00 - £359.00
Specifications
| Antigen | IL4R |
|---|---|
| Clone | 102 |
| Concentration | 100 μg/mL |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30019439
|
Invitrogen™
MA552206 |
20 μL |
£124.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30019869
![]() |
Invitrogen™
MA552205 |
100 μL |
£359.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL4R Recombinant Monoclonal antibody specifically detects IL4R in Human samples. It is validated for ELISASpecifications
| IL4R | |
| 100 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| 582J2.1; CD124; il-4 receptor; IL-4 receptor alpha chain; IL-4 receptor subunit alpha; IL-4-binding protein; IL4-BP; Il4r; IL4R nirs variant 1; IL-4R subunit alpha; IL4RA; IL-4RA; IL-4R-alpha; interleukin 4 receptor; interleukin 4 receptor, alpha; interleukin-4 receptor alpha chain; interleukin-4 receptor subunit alpha; Soluble IL-4 receptor subunit alpha; Soluble interleukin-4 receptor subunit alpha | |
| Recombinant Human IL4R protein (Met 26-His232). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| IL4R |
| 102 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P24394 | |
| 3566 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title