missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL4I1 Polyclonal Antibody

IL4I1 Polyclonal Antibody
Brand: Thermo Scientific PA5115418
This item is not returnable.
View return policy
Description
IL4I1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (PFA)
Specifications
| IL4I1 | |
| Polyclonal | |
| Unconjugated | |
| IL4I1 | |
| FIG1; Fig-1 protein; Fig1-ps; hFIG1; IL4I1; IL4I1_2; IL4-induced protein 1; interleukin 4 induced 1; interleukin four induced 1; interleukin-4-induced protein 1; interleukin-four induced gene 1; interleukin-four induced gene-1 protein; interleukin-four induced protein-1; LAAO; L-amino acid oxidase; L-amino-acid oxidase; LAO; LOC100360621; mFIG1; Protein Fig-1; rCG53598-like; UNQ636/PRO1265 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 100360621, 259307 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q96RQ9 | |
| IL4I1 | |
| A synthesized peptide derived from human IL4I1(Accession Q96RQ9), corresponding to amino acid residues P486-S536. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction