missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ IL4 (Interleukin 4); Recombinant Human Protein

Product Code. 10680861
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10680861 100 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10680861 Supplier Thermo Scientific™ Supplier No. CTP0041

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

IL4 (Interleukin 4); Recombinant Human Protein

IL4 promotes the proliferation and differentiation of activated B cells. In addition, IL4 up-regulates class II MHC antigen expression and IgE receptors. ED50 = 0.05 to 0.4ng/mL (Specific Activity: 2.0 x 107 to 2.5 x 106 units/mg). The biological activity is determined by measuring the dose-dependent proliferation of human TF-1 cells.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Primary Cell Culture, Cell Culture, Mammalian Cell Culture, Dendritic Cell Culture, Mesenchymal Stem Cell Culture, Stem Cell Research, Stem Cell Culture, Hematopoietic Stem Cell Culture
Formulation Carrier-Free
Molecular Weight (g/mol) 14.9 kDa
Purification Method Gel-Purified
Quantity 100 μg
Storage Requirements Lyophilized protein can be stored at 2°C to 8°C dessicated. Once reconstituted, store in aliquots at ≤20°C. Avoid multiple freeze/thaw cycles.
Regulatory Status RUO - research use only
Structural Form Full Length, Recombinant, Ligand
Endotoxin Concentration <0.1 ng/μg
Gene Alias IL-4 (Interleukin 4)
Cross Reactivity Human
Species Human
Recombinant Recombinant
Expression System E. coli
Protein Family Cytokines & Receptors
Protein Subtype Interleukins
Purity or Quality Grade 95 %
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.