missing translation for 'onlineSavingsMsg'
Learn More

IL2RA Mouse anti-Human, Clone: TP1/6 (Biotin), Abnova™

Product Code. 16052396
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16052396 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16052396 Supplier Abnova Supplier No. MAB13855.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human IL2RA.

  • High specificity
  • High reactivity

Specifications

Antigen IL2RA
Applications Flow Cytometry
Classification Monoclonal
Clone TP1/6
Conjugate Biotin
Description Mouse monoclonal antibody raised against human IL2RA.
Dilution Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
Gene Accession No. P01589
Gene Alias CD25/IDDM10/IL2R/TCGFR
Gene Symbols IL2RA
Host Species Mouse
Immunogen PHA activated human T cells.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3559
Target Species Human
Content And Storage Store in the dark at 4°C. Avoid prolonged exposure to light.
Form Liquid
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.