Learn More
Invitrogen™ IL24 Recombinant Rabbit Monoclonal Antibody (101)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA530578
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human IL-24/MDA-7.
Interleukin 24 (IL-24)/melanoma differentiation-associated gene-7 (mda-7) encodes a secreted protein that exhibits significant homology to the interleukin 10 (IL-10) family of cytokines. The human IL-24 is secreted by activated peripheral blood mononuclear cells and binds two heterodimeric receptors, IL-22R1/IL-20R2 and IL-20R1/IL-20R2 resulting in the activation of JAK/STAT signaling pathways. Mda-7/IL-24, selectively induces growth suppression, apoptosis and radiosensitization in malignant gliomas in a p53-independent manner.
Specifications
| IL24 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q13007 | |
| IL24 | |
| Recombinant Human IL-24/MDA-7 Protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 101 | |
| Unconjugated | |
| IL24 | |
| C49A; Cytokine-like protein Mob-5; FISP; IL 24; IL10B; Il24; IL-24; IL-4-induced secreted protein; Interleukin; interleukin 24; interleukin-24; Mda7; MDA-7; melanocyte-associated Mda-7; melanoma differentiation-associated gene 7 protein; Mob5; ST16; suppression of tumorigenicity 16 (melanoma differentiation); Suppression of tumorigenicity 16 protein; Th2-specific cytokine FISP | |
| Rabbit | |
| Protein A | |
| RUO | |
| 11009 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.