missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL23R Recombinant Rabbit Monoclonal Antibody (3Y7B9)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558205

 View more versions of this product

Product Code. 30283167

  • £147.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: GITSINCSGD MWVEPGEIFQ MGMNVSIYCQ EALKHCRPRN LYFYKNGFKE EFDITRINRT TARIWYKGFS EPHAYMHCTA ECPGHFQETL ICGKDISSGH PPDAPSNLTC VIYEYSGNMT CTWNTGKPTY IDTKYIVHVK SLETEEEQQY LASSYVKIST DSLQGSRKYL VWVQAVNSLG MENSQQLHVH LDDIVIPSAS IISRAETTND TVPKTIVYWK SKTMIEKVFC EMRYKTTTNQ TWSVKEFDAN FTYVQ.

IL-23R is a 629 aa, novel subunit of the receptor for cytokine IL23A/IL23 which pairs with the receptor molecule IL12RB1/IL12R beta 1and thus take part in IL23A signaling. Signal transduction analysis confirms that this protein associates constitutively with Janus kinase 2 (JAK2), and also binds to transcription activator STAT3 in a ligand-dependent manner. Alternative splicing results in six spliced isoforms of IL-23R. Reports suggest that IL-23R plays an important role in psoriasis pathogenesis and is also a potential therapeutic target in inflammatory bowel disease.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL23R
Recombinant Monoclonal
0.22 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Q8K4B4
Il23r
Recombinant fusion protein containing a sequence corresponding to amino acids 24-315 of mouse IL-23R (NP_653131.3).
20 μL
Primary
Mouse
Antibody
IgG
ELISA, Western Blot
3Y7B9
Unconjugated
Il23r
IBD17; IL 23R; IL-23 receptor; IL23R; IL-23R; interleukin 23 receptor; interleukin 23 receptor-like; interleukin-23 receptor
Rabbit
Protein A
RUO
209590
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.