Learn More
IL22RA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13462-1-AP-20UL
 View more versions of this product
Product Code. 16861694
  • £135.00 / 20µL
Description
A novel cytokine, designated IL-TIF for IL-10 related T cell-derived inducible factor and IL-22, was recently identified. The receptor for IL-22 (IL-22R, also termed CRF2-9 and IL-TIF-R1 chain) is a new member of the class II cytokine receptor family. IL-22R forms a complex with IL-10 receptor beta chain and mediates IL-22 signaling. IL-22 and its receptor activate JAK-STAT signaling pathway. IL22R is expressed in normal liver and kidney and their cell lines HepG2 and TK-10. A soluble form of IL-22 receptor, also termed IL-22 binding protein (IL-22BP) and IL-22 receptor-alpha 2 (IL-22RA2), was identified very recently. IL-22BP prevents binding of IL-22 to the functional cell surface IL-22R complex and neutralizes IL-22 activity. LPS induces IL-22 expression, which indicates the role of IL-22 in inflammatory response.
Specifications
Specifications
IL22RA1
Polyclonal
Unconjugated
IL22RA1
CRF2 9, IL 22 receptor subunit alpha 1, IL 22R alpha 1, IL 22RA1, IL22R, IL22R1, IL22RA1, ZcytoR11
Rabbit
Antigen Affinity Chromatography
RUO
230828, 362629, 58985
-20°C
Liquid
Flow Cytometry, Immunofluorescence, Immunocytochemistry, Western Blot
0.53 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q80XZ4, Q8N6P7
Il22ra1
IL22RA1 Fusion Protein Ag4056
20 μL
Primary
Rat, Human, Mouse