missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL21R Monoclonal Antibody (1C3G9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA548744
This item is not returnable.
View return policy
Description
IL21R Monoclonal Antibody for Western Blot, ICC/IF, Flow, ELISA
The protein encoded by this gene is a cytokine receptor for interleukin 21 (IL21). It belongs to the type I cytokine receptors, and has been shown to form a heterodimeric receptor complex with the common gamma-chain, a receptor subunit also shared by the receptors for interleukin 2 (IL2) and interleukin 5 (IL5). This receptor transduces the growth promoting signal of IL21, and is important for the proliferation and differentiation of T cells, B cells, and natural killer (NK) cells. The ligand binding of this receptor leads to the activation of multiple downstream signaling molecules, including JAK1, JAK3, STAT1, and STAT3. Knockout studies of a similar gene in mouse suggest a role for this gene in regulating immunoglobulin production. Three alternatively spliced transcript variants encoding the same protein have been described.
Specifications
| IL21R | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q9HBE5 | |
| Il21r | |
| Purified recombinant fragment of human CD360 (AA: extra 20-232) expressed in HEK293-6e cells supernatant. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry, Western Blot | |
| 1C3G9 | |
| Unconjugated | |
| Il21r | |
| CD360; IL-21 receptor; Il21r; IL-21R; interleukin 21 receptor; interleukin-21 receptor; LR-beta; lymphocyte receptor beta; MGC10967; NILR; novel cytokine receptor 8; Novel interleukin receptor; NR8; UNQ3121/PRO10273 | |
| Mouse | |
| Protein G | |
| RUO | |
| 50615 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction