missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL20RA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA141167
This item is not returnable.
View return policy
Description
This antibody is predicted to react with chimpanzee and rat samples based on sequence homology. Suggested positive control: A375. The amino acid sequence used as immunogen is 100% homologous in human and chimp, and 82% homologous in mouse and rat.
The protein encoded by this gene is a receptor for interleukin 20 (IL20), a cytokine that may be involved in epidermal function. The receptor of IL20 is a heterodimeric receptor complex consisting of this protein and interleukin 20 receptor beta (IL20B). This gene and IL20B are highly expressed in skin. The expression of both genes is found to be upregulated in Psoriasis.
Specifications
| IL20RA | |
| Polyclonal | |
| Unconjugated | |
| Il20ra | |
| class II cytokine receptor ZCYTOR7; CRF2-8; Cytokine receptor class-II member 8; Cytokine receptor family 2 member 8; E230031K19; IL-20 receptor subunit alpha; IL-20R1; IL20RA; IL-20RA; IL-20R-alpha; interleukin 20 receptor alpha subunit; interleukin 20 receptor subunit alpha; interleukin 20 receptor, alpha; interleukin-20 receptor I; interleukin-20 receptor subunit alpha; PRO1315; UNQ681; UNQ681/PRO1315; ZcytoR7 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 237313, 308716, 53832 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 1.0 mg/mL | |
| PBS with 0.2% gelatin and 0.05% sodium azide | |
| Q6PHB0, Q9UHF4 | |
| Il20ra | |
| Amino acids 159-175 of human IL-20 receptor alpha protein. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction