missing translation for 'onlineSavingsMsg'
Learn More

IL1RAPL2 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18395672 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18395672 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18395672 Supplier Novus Biologicals Supplier No. NBP298932AF405

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IL1RAPL2 Polyclonal specifically detects IL1RAPL2 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL1RAPL2
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50 mM Sodium Borate
Gene Alias IL-1R-9, IL1R9IL-1 receptor accessory protein-like 2, IL-1R9IL-1RAPL-2, IL-1-RAPL-2, IL1RAPL-2IL1RAPL-2-related protein, interleukin 1 receptor 9, interleukin 1 receptor accessory protein-like 2, Three immunoglobulin domain-containing IL-1 receptor-related 1, TIGIRR-1Interleukin-1 receptor 9, X-linked interleukin-1 receptor accessory protein-like 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant IL1RAPL2 (NP_059112.1; Met 1-Glu 356).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26280
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.