Learn More
Invitrogen™ IL1RAPL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596244
Description
Immunogen sequence: SISLTVGEND TGLCYNSKMK YFEKAELSKS KEISCRDIED FLLPTREPEI LWYKECRTKT WRPSIVFKRD TLLIREVRED D; Positive Samples: A-431, Mouse brain, Mouse heart, Rat brain, Rat heart; Cellular Location: Cell membrane, Cell projection, Cytoplasm, Single-pass type I membrane protein, axon, dendrite.
The protein encoded by this gene is a member of the interleukin 1 receptor family and is similar to the interleukin 1 accessory proteins. This protein has an N-terminal signal peptide, three extracellular immunoglobulin Ig-like domains, a transmembrane domain, an intracellular Toll/IL-1R domain, and a long C-terminal tail which interacts with multiple signaling molecules. This gene is located at a region on chromosome X that is associated with a non-syndromic form of X-linked intellectual disability. Deletions and mutations in this gene were found in patients with intellectual disability. This gene is expressed at a high level in post-natal brain structures involved in the hippocampal memory system, which suggests a specialized role in the physiological processes underlying memory and learning abilities, and plays a role in synapse formation and stabilization.
Specifications
| IL1RAPL1 | |
| Polyclonal | |
| Unconjugated | |
| Il1rapl1 | |
| 6330532G10Rik; C030039N24; IL1R8; Il1rapl; Il1rapl1; IL-1-RAPL-1; IL-1RAPL-1; IL1RAPL-1; interleukin 1 receptor accessory protein like 1; interleukin 1 receptor accessory protein-like 1; interleukin 1 receptor-8; interleukin-1 receptor accessory protein-like 1; mental retardation, X-linked 10; MRX10; MRX21; MRX34; Oligophrenin-4; OPHN4; Three immunoglobulin domain-containing IL-1 receptor-related 2; TIGIRR-2; X-linked interleukin-1 receptor accessory protein-like 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11141, 317553, 331461 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.07 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P59823, P59824, Q9NZN1 | |
| Il1rapl1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 130-210 of human IL1RAPL1 (NP_0550861). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.