missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL1RAPL1 Mouse anti-Human, Clone: B-R58, Novus Biologicals™
Mouse Monoclonal Antibody
£324.00 - £600.00
Specifications
| Antigen | IL1RAPL1 |
|---|---|
| Clone | B-R58 |
| Concentration | 1 mg/ml |
| Dilution | Flow Cytometry, ELISA |
| Applications | Flow Cytometry, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18331637
|
Novus Biologicals
NBP3-18111-200UL |
200 μg |
£324.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18337576
|
Novus Biologicals
NBP3-18111-500UL |
500 μg |
£600.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL1RAPL1 Monoclonal antibody specifically detects IL1RAPL1 in Human samples. It is validated for Flow Cytometry, ELISASpecifications
| IL1RAPL1 | |
| 1 mg/ml | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| IL1R8IL1RAPL, IL-1-RAPL-1, IL-1RAPL-1, IL1RAPL-1, interleukin 1 receptor accessory protein-like 1, interleukin 1 receptor-8, interleukin-1 receptor accessory protein-like 1, mental retardation, X-linked 10, MRX10, MRX21, MRX34, Oligophrenin-4, OPHN4mental retardation, X-linked 21, Three immunoglobulin domain-containing IL-1 receptor-related 2, TIGIRR-2mental retardation, X-linked 34, X-linked interleukin-1 receptor accessory protein-like 1 | |
| Recombinant human IL1RAPL1. Hybridoma: Myeloma X63/AG.8653 x Balb/c spleen cells | |
| Primary | |
| Store at 4C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles. |
| B-R58 | |
| Flow Cytometry, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS. Sterile-filtered through 0.22 um. | |
| 11141 | |
| IgG1 κ | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title