missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL1RAPL1 Antibody (2H3C12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£435.00
Specifications
| Antigen | IL1RAPL1 |
|---|---|
| Clone | 2H3C12 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry |
Description
IL1RAPL1 Monoclonal specifically detects IL1RAPL1 in Human samples. It is validated for Western Blot, ELISA, CyTOF-ready.Specifications
| IL1RAPL1 | |
| 1 mg/mL | |
| Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| IL1R8IL1RAPL, IL-1-RAPL-1, IL-1RAPL-1, IL1RAPL-1, interleukin 1 receptor accessory protein-like 1, interleukin 1 receptor-8, interleukin-1 receptor accessory protein-like 1, mental retardation, X-linked 10, MRX10, MRX21, MRX34, Oligophrenin-4, OPHN4mental retardation, X-linked 21, Three immunoglobulin domain-containing IL-1 receptor-related 2, TIGIRR-2mental retardation, X-linked 34, X-linked interleukin-1 receptor accessory protein-like 1 | |
| IL1RAPL1 | |
| IgG1 | |
| Protein A or G purified |
| 2H3C12 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 11141 | |
| Purified recombinant fragment of human IL1RAPL1 (AA: 541-694) expressed in E. Coli. | |
| Primary | |
| 80 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title