Learn More
IL1RA Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10844-1-AP-20UL
 View more versions of this product
Product Code. 16837503
  • £135.00 / 20µL
Description
IL1RA (IL1RN) protein is a cytokine receptor that belongs to the interleukin 1 receptor family. IL1RA is a receptor for interleukin alpha (IL1A), interleukin beta (IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA). It is an important mediator involved in many cytokine induced immune and inflammatory responses. This gene along with interleukin 1 receptor, type II (IL1R2), interleukin 1 receptor-like 2 (IL1RL2), and interleukin 1 receptor-like 1 (IL1RL1) form a cytokine receptor gene cluster in a region mapped to chromosome 2q12.
Specifications
Specifications
IL1RA
Polyclonal
Unconjugated
IL1RN
Anakinra, ICIL 1RA, IL 1ra, IL 1ra3, IL 1RN, IL1 inhibitor, IL1F3, IL1RA, IL1RN, IRAP
Rabbit
Antigen Affinity Chromatography
RUO
3557
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P18510
IL1RN
IL1RA Fusion Protein Ag1277
20 μL