missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL1RA Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10844-1-AP-150UL
This item is not returnable.
View return policy
Description
IL1RA (IL1RN) protein is a cytokine receptor that belongs to the interleukin 1 receptor family. IL1RA is a receptor for interleukin alpha (IL1A), interleukin beta (IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA). It is an important mediator involved in many cytokine induced immune and inflammatory responses. This gene along with interleukin 1 receptor, type II (IL1R2), interleukin 1 receptor-like 2 (IL1RL2), and interleukin 1 receptor-like 1 (IL1RL1) form a cytokine receptor gene cluster in a region mapped to chromosome 2q12.Specifications
| IL1RA | |
| Polyclonal | |
| Unconjugated | |
| IL1RN | |
| Anakinra, ICIL 1RA, IL 1ra, IL 1ra3, IL 1RN, IL1 inhibitor, IL1F3, IL1RA, IL1RN, IRAP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3557 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P18510 | |
| IL1RN | |
| IL1RA Fusion Protein Ag1277 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction