Learn More
Invitrogen™ IL1R2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547759
Description
In direct ELISAs, approximately 30% cross-reactivity with recombinant humanIL-1 RII is observed and less than 1% cross-reactivity with recombinant mouse (rm) IL-1 RI, recombinant rat IL-1 Rrp2, rmIL-18 R, andrmIL-18 R beta is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
The protein encoded by this gene is a cytokine receptor that belongs to the interleukin 1 receptor family. This protein binds interleukin alpha (IL1A), interleukin beta (IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA), and acts as a decoy receptor that inhibits the activity of its ligands. Interleukin 4 (IL4) is reported to antagonize the activity of interleukin 1 by inducing the expression and release of this cytokine. This gene and three other genes form a cytokine receptor gene cluster on chromosome 2q12. Two alternatively spliced transcript variants encoding the same protein have been reported.
Specifications
| IL1R2 | |
| Polyclonal | |
| Unconjugated | |
| Il1r2 | |
| antigen CDw121b; CD121 antigen-like family member B; CD121b; CDw121b; IL-1 receptor beta chain; IL-1 type II receptor; Il1r2; Il-1r2; IL-1R-2; Il1r-2; IL1R2c; IL1RB; IL-1R-beta; IL-1RT2; IL-1RT-2; ILR2; interleukin 1 receptor type 2; interleukin 1 receptor type II variant 3; interleukin 1 receptor, type II; interleukin-1 receptor beta; Interleukin-1 receptor type 2; Interleukin-1 receptor type 2, membrane form; Interleukin-1 receptor type 2, soluble form; Interleukin-1 receptor type II; mIL-1R2; sIL-1R2; type II interleukin-1 receptor, beta | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 16178 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Frozen), Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P27931 | |
| Il1r2 | |
| Sf 21-derived recombinant mouse IL-1 RII Phe14-Glu355 (predicted). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.