missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL1R2 Mouse anti-Human, Mouse, Clone: 3H4H4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60262-1-IG-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a cytokine receptor that belongs to the interleukin 1 receptor family. This protein binds interleukin alpha (IL1A), interleukin beta (IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA), and acts as a decoy receptor that inhibits the activity of its ligands. Interleukin 4 (IL4) is reported to antagonize the activity of interleukin 1 by inducing the expression and release of this cytokine. This gene and three other genes form a cytokine receptor gene cluster on chromosome 2q12. Two alternatively spliced transcript variants encoding the same protein have been reported.Specifications
| IL1R2 | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P27930, P27931 | |
| Il1r2 | |
| IL1R2 Fusion Protein Ag4495 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 3H4H4 | |
| Unconjugated | |
| IL1R2 | |
| CD121b, CDw121b, IL 1R 2, IL 1R beta, IL 1RT 2, IL 1RT2, IL1 Receptor 2, IL-1 Receptor 2, IL1 Receptor II, IL-1 Receptor II, IL1R2, IL1RB, IL-1RII, Interleukin 1 receptor beta, Interleukin 1 receptor type 2, Interleukin 1 receptor type II | |
| Mouse | |
| Protein A | |
| RUO | |
| 16178, 7850 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction