missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL1F5 Monoclonal Antibody (04), Invitrogen™

Mouse Monoclonal Antibody
£219.00 - £384.00
Specifications
| Antigen | IL1F5 |
|---|---|
| Clone | 04 |
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17779863
|
Invitrogen™
MA543519 |
50 μL |
£219.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17709533
![]() |
Invitrogen™
MA542975 |
100 μL |
£384.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specificity: Human IL-1F5
The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine was shown to specifically inhibit the activation of NF-kappaB induced by interleukin 1 family, member 6 (IL1F6). This gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. Two alternatively spliced transcript variants encoding the same protein have been reported.Specifications
| IL1F5 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| AI413231; FIL1; FIL1 delta; FIL1(DELTA); FIL1D; Fil1delta; IL-1 delta; IL-1 related protein 3; Il1f5; IL-1F5; IL1F5 (Canonical product IL-1F5a); IL-1F5 (IL-1HY1, FIL1-delta, IL-1RP3, IL-1L1, IL-1-delta); Il1h3; Il-1h3; IL1HY1; IL-1HY1; IL1L1; IL-1L1; IL-1ra homolog 1; IL-1-related protein 3; IL1RP3; IL-1RP3; IL36RA; IL-36Ra; IL36RN; Interleukin; interleukin 1 family, member 5; interleukin 1 family, member 5 (delta); interleukin 1 receptor antagonist homolog 1; interleukin 36 receptor antagonist; interleukin-1 delta; Interleukin-1 family member 5; interleukin-1 family member 5; interleukin-36 receptor antagonist protein; interleukin-1 homolog 3; Interleukin-1 HY1; interleukin-1 receptor antagonist homolog 1; Interleukin-1-like protein 1; interleukin-36 receptor antagonist protein; MGC29840; PSORP; PSORS14; UNQ1896/PRO4342 | |
| IL36RN | |
| IgG1 | |
| Protein A | |
| Antibody |
| 04 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9UBH0 | |
| 26525 | |
| Recombinant Human IL-1F5 Protein. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| IL36RN |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title