missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL1F10 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16677314
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16677314 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16677314 Supplier Invitrogen™ Supplier No. PA5100834

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IL1FA.

The cytokine IL-1 is responsible for initiating a variety of activities through the activation of transcription factors, NF-kappa B and AP-1, thereby promoting host response to injury or infection. IL-1F10 (Interleukin-1 family member 10), also known as FIL1T, IL1HY2 or FKSG75, is highly expressed in the skin, spleen and proliferating B-cells of the tonsil. IL-1F10 binds soluble IL-1 receptor type 1, and is one of nine Interleukin 1 families clustered on chromosome 2, where it is thought to participate in the regulation of adapted and innate immune responses. IL-1F10 and IL-1RN polymorphisms may play an important role in the susceptibility to developing rheumatoid arthritis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL1F10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IL1F10
Gene Accession No. Q8R459, Q8WWZ1
Gene Alias family of interleukin 1-theta; FIL1 theta; FIL1T; FIL1-theta; FKSG75; IL-1 theta; Il1f10; IL-1F10; IL-1F10 (canonical form IL-1F10a); IL1HY2; IL-1HY2; IL1-theta; IL38; IL-38; Interleukin; interleukin 1 family member 10 (theta); interleukin 1 family, member 10; interleukin 1 family, member 10 (theta); interleukin 1 family, member 7; interleukin-1 family member 10; Interleukin-1 HY2; interleukin-1 receptor antagonist FKSG75; interleukin-1 receptor antagonist-like FIL1 theta; Interleukin-1 theta; Interleukin-38; UNQ6119/PRO20041
Gene Symbols IL1F10
Host Species Rabbit
Immunogen A synthesized peptide derived from human IL1F10(Accession Q8WWZ1), corresponding to amino acid residues S104-W152.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 215274, 84639
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.