missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL1F10 Mouse anti-Human, Clone: 5B2F5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60319-1-IG-150UL
This item is not returnable.
View return policy
Description
The cytokine IL-1 is responsible for initiating a variety of activities through the activation of transcription factors, NF-kappa B and AP-1, thereby promoting host response to injury or infection. IL-1F10 (Interleukin-1 family member 10), also known as FIL1T, IL1HY2 or FKSG75, is highly expressed in the skin, spleen and proliferating B-cells of the tonsil. IL-1F10 binds soluble IL-1 receptor type 1, and is one of nine Interleukin 1 families clustered on chromosome 2, where it is thought to participate in the regulation of adapted and innate immune responses. IL-1F10 and IL-1RN polymorphisms may play an important role in the susceptibility to developing rheumatoid arthritis.Specifications
| IL1F10 | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8WWZ1 | |
| IL1F10 | |
| IL1F10 Fusion Protein Ag17250 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 5B2F5 | |
| Unconjugated | |
| IL1F10 | |
| FIL1 theta, FIL1T, FKSG75, IL 1 theta, IL 1F10, IL 1HY2, IL1 theta, IL1F10, IL1HY2, Interleukin 1 family member 10, Interleukin 1 HY2, Interleukin 1 theta, UNQ6119/PRO20041 | |
| Mouse | |
| Protein G | |
| RUO | |
| 84639 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering