missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL18RAP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580721
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human IL18RAP.
The protein encoded by this gene is an accessory subunit of the heterodimeric receptor for IL18. This protein enhances the IL18 binding activity of IL18R1 (IL1RRP), a ligand binding subunit of IL18 receptor. The coexpression of IL18R1 and this protein is required for the activation of NF-kappaB and MAPK8 (JNK) in response to IL18.
Specifications
| IL18RAP | |
| Polyclonal | |
| Unconjugated | |
| IL18RAP | |
| Accessory protein-like; ACPL; AcPL accessory protein-like); CD218 antigen-like family member B; CD218b; CDw218b; cluster of differentiation w218b; IL-18 RA; IL-18 receptor accessory protein; IL-18 receptor beta; IL-18RAcP; IL18RAP; IL18RB; IL-18Rbeta; IL-18R-beta; IL-1R accessory protein-like; IL1R7; IL-1R7; IL-1R-7; IL-1RAcPL; interleukin 18 receptor accessory protein; interleukin-1 receptor 7; Interleukin-18 receptor accessory protein; Interleukin-18 receptor accessory protein-like; Interleukin-18 receptor beta | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 8807 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| Conc. not determined | |
| PBS with no preservative | |
| O95256 | |
| IL18RAP | |
| Recombinant Human IL18RAP Protein (Met1-Gly357). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction