missing translation for 'onlineSavingsMsg'
Learn More

IL18R1 Mouse anti-Human, Clone: B-E43, Novus Biologicals™

Product Code. 18329571
Change view
Click to view available options
Quantity:
200 μg
500 μg
Unit Size:
200µL
500µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18329571 500 μg 500µL
18008625 200 μg 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18329571 Supplier Bio-Techne Supplier No. NBP314586500UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL18R1 Monoclonal antibody specifically detects IL18R1 in Human samples. It is validated for Flow Cytometry, ELISA, Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL18R1
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone B-E43
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry, ELISA, Functional
Formulation PBS. Sterile-filtered through 0.22 um.
Gene Alias CD218 antigen-like family member A, CD218a, CD218a antigen, CDw218a, cytokine receptor, IL-18R1, IL-18R-1, IL1R-rp, IL1RRPIL1 receptor-related protein, IL-1RrpIL18RA, interleukin 18 receptor 1, interleukin-18 receptor 1
Host Species Mouse
Immunogen Recombinant human IL18R1
Purification Method Affinity purified
Quantity 500 μg
Regulatory Status RUO
Research Discipline CD Markers
Primary or Secondary Primary
Gene ID (Entrez) 8809
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.