missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL18R1 Monoclonal Antibody (05)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA530547
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human IL18R1.
IL18R1 is a cytokine receptor that belongs to the interleukin 1 receptor family. This receptor specifically binds interleukin 18 (IL18), and is essential for IL18 mediated signal transduction. IFN-alpha and IL12 are reported to induce the expression of this receptor in NK and T cells. The gene along with four other members of the interleukin 1 receptor family, including IL1R2, IL1R1, ILRL2 (IL-1Rrp2), and IL1RL1 (T1/ST2), form a gene cluster on chromosome 2q.
Specifications
| IL18R1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q13478 | |
| Il18r1 | |
| Recombinant Human IL18R1 Protein (Met1-Arg329). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 05 | |
| Unconjugated | |
| Il18r1 | |
| CD218 antigen-like family member A; CD218a; CDw218a; cytokine receptor; IL1 receptor-related protein; Il18r1; IL-18R1; IL-18R-1; IL18RA; Il18ralpha; IL-18R-alpha; IL18Ralpha2; IL1RRP; IL-1Rrp; IL1R-rp; interleukin 1 receptor related protein; interleukin 18 receptor 1; Interleukin-18; interleukin-18 receptor 1; Interleukin-18 receptor alpha | |
| Mouse | |
| Protein A | |
| RUO | |
| 8809 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction