Learn More
Invitrogen™ IL18R1 Monoclonal Antibody (112624)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA523855
Description
In direct ELISAs, this antibody does not cross-react with recombinant mouse (rm) IL-1 RI, rmIL-1 RII, rhIL-1 R3, recombinant human (rh) IL-1 R4, rhIL-1 R5, recombinant rat IL-1 R6, rmIL-1 R7, rhIL-1 R8, rhIL-1 R9, or rmSIGIRR. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
IL18R1 is a cytokine receptor that belongs to the interleukin 1 receptor family. This receptor specifically binds interleukin 18 (IL18), and is essential for IL18 mediated signal transduction. IFN-alpha and IL12 are reported to induce the expression of this receptor in NK and T cells. The gene along with four other members of the interleukin 1 receptor family, including IL1R2, IL1R1, ILRL2 (IL-1Rrp2), and IL1RL1 (T1/ST2), form a gene cluster on chromosome 2q.
Specifications
| IL18R1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q61098 | |
| Il18r1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-18 R alpha Ser19-Gly326. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Neutralization, Western Blot | |
| 112624 | |
| Unconjugated | |
| Il18r1 | |
| CD218 antigen-like family member A; CD218a; CDw218a; cytokine receptor; IL1 receptor-related protein; Il18r1; IL-18R1; IL-18R-1; IL18RA; Il18ralpha; IL-18R-alpha; IL18Ralpha2; IL1RRP; IL-1Rrp; IL1R-rp; interleukin 1 receptor related protein; interleukin 18 receptor 1; Interleukin-18; interleukin-18 receptor 1; Interleukin-18 receptor alpha | |
| Rat | |
| Protein A/G | |
| RUO | |
| 16182 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.