missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IL18R1 Antibody Pair [HRP]
Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IL18R1.
Specifications
Specifications
| Assay Range | 7.8 pg/mL |
| Assay Sensitivity | 7.81-500 pg/mL |
| Assay | Sandwich ELISA |
| Conjugate | HRP |
| Product Type | Antibody Pairs |
| Specificity | This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IL18R1. |
| Target Species | Human |
| For Use With (Application) | Sandwich ELISA |
| Gene Alias | CD218 antigen-like family member A, CD218a, CD218a antigen, CDw218a, cytokine receptor, IL-18R1, IL-18R-1, IL1R-rp, IL1RRPIL1 receptor-related protein, IL-1RrpIL18RA, interleukin 18 receptor 1, interleukin-18 receptor 1 |
| Gene ID (Entrez) | 8809 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction