missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL18R1 Antibody (MM0380-8H22), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11679PCP
This item is not returnable.
View return policy
Description
IL18R1 Monoclonal antibody specifically detects IL18R1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Neutralization
Specifications
| IL18R1 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Neutralization | |
| MM0380-8H22 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize | |
| CD218 antigen-like family member A, CD218a, CD218a antigen, CDw218a, cytokine receptor, IL-18R1, IL-18R-1, IL1R-rp, IL1RRPIL1 receptor-related protein, IL-1RrpIL18RA, interleukin 18 receptor 1, interleukin-18 receptor 1 | |
| Human recombinant IL18R1 (extracellular domain) | |
| 0.1 mL | |
| CD Markers | |
| 8809 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction