Learn More
IL18R1 Antibody (MM0380-8H22), mFluor Violet 450 SE, Novus Biologicals™
Product Code. 18400618 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
Product Code. unitSize
18400618 0.1mL
1 options
Product Code. 18400618 Supplier Novus Biologicals Supplier No. NBP211679MFV450
Mouse Monoclonal Antibody
IL18R1 Monoclonal antibody specifically detects IL18R1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Neutralization
TRUSTED_SUSTAINABILITY
Specifications
Applications Immunohistochemistry (Paraffin), Neutralization