missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL18R1 Antibody (11), DyLight 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89797UV
This item is not returnable.
View return policy
Description
IL18R1 Monoclonal specifically detects IL18R1 in Human samples. It is validated for ELISA.
Specifications
| IL18R1 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 8809 | |
| Human | |
| Purified |
| ELISA | |
| 11 | |
| ELISA | |
| CD218 antigen-like family member A, CD218a, CD218a antigen, CDw218a, cytokine receptor, IL-18R1, IL-18R-1, IL1R-rp, IL1RRPIL1 receptor-related protein, IL-1RrpIL18RA, interleukin 18 receptor 1, interleukin-18 receptor 1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human IL18R1 (NP_003846.1; Met 1-Arg 329). | |
| 0.1 mL | |
| Primary | |
| No cross-reactivity in ELISA with: Mouse IL18R1/ IL1RRP/ CD218a Mouse SIGIRR Human IL1R1/ CD121a Human IL1R2/ IL1RB/ CD121b Human IL1R3/ IL1RAP Human IL1R8/ IL1RAPL1 Human IL1RL1/ IL1R4/ ST2 Human IL18RAP/ IL1R7 Human IL1R9/ IL1RAPL2 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction